MAT100860154 Matrix Scientific

4-m-Tolyl-tetrahydro-pyran-4-carboxylic acid

  • MDL number MFCD08445815
  • Cas Number : 889940-10-7
  • Molecular Weight : 220.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin US
Product Link https://www.matrixscientific.com/021219.html

Safety Information

hcodes Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart