MAT100860165 Matrix Scientific

7-Methylsulfanyl-2-oxo-1,2-dihydro-quinoline-3-carbaldehyde

  • MDL number MFCD08445792
  • Cas Number : 873300-63-1
  • Molecular Weight : 219.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/021230.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/021230.html
Availability details for
Quantity
  • Availability
  • Add to Cart