MAT100860219 Matrix Scientific

5-tert-Butyl-4-morpholin-4-ylmethyl-furan-2-carboxylic acid hydrochloride

  • MDL number MFCD03997883
  • Cas Number : 1052420-48-0
  • Molecular Weight : 303.79

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/021285.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/021285.html
Availability details for
Quantity
  • Availability
  • Add to Cart