MAT100860224 Matrix Scientific

Benzyl-ethyl-piperidin-4-yl-amine

  • MDL number MFCD03274735
  • Cas Number : 76167-64-1
  • Molecular Weight : 218.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Boiling point 133-135�15 Torr
Product Link http://www.matrixscientific.com/021290.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/021290.html
Availability details for
Quantity
  • Availability
  • Add to Cart