MAT100860231 Matrix Scientific

3-(Piperidin-4-yloxy)-pyridine

  • MDL number MFCD08059774
  • Cas Number : 310881-48-2
  • Molecular Weight : 178.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/021297.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/021297.html
Availability details for
Quantity
  • Availability
  • Add to Cart