MAT100860237 Matrix Scientific

2,2-Dimethyl-2,3-dihydro-benzofuran-5-carbaldehyde

  • MDL number MFCD08445737
  • Cas Number : 38002-92-5
  • Molecular Weight : 176.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/021303.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/021303.html
Availability details for
Quantity
  • Availability
  • Add to Cart