MAT100860247 Matrix Scientific

4-(2-Fluoro-phenyl)-tetrahydro-pyran-4-carboxylic acid

  • MDL number MFCD08445765
  • Cas Number : 889939-77-9
  • Molecular Weight : 224.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/021313.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/021313.html
Availability details for
Quantity
  • Availability
  • Add to Cart