MAT100860257 Matrix Scientific

(2,2,6,6-Tetramethyl-1,2,3,6-tetrahydro-pyridin-4-yl)-acetic acid

  • MDL number MFCD03767237
  • Cas Number : 88461-91-0
  • Molecular Weight : 197.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/021323.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/021323.html
Availability details for
Quantity
  • Availability
  • Add to Cart