MAT100860289 Matrix Scientific

1-(2-Isopropoxy-ethyl)-piperazine

  • MDL number MFCD08445769
  • Cas Number : 889939-82-6
  • Molecular Weight : 172.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/021357.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/021357.html
Availability details for
Quantity
  • Availability
  • Add to Cart