MAT100860301 Matrix Scientific

5-Bromo-6-chloronicotinic acid

  • MDL number MFCD01927098
  • Cas Number : 29241-62-1
  • Molecular Weight : 236.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 166-168
Purity 98
Country of Origin USA
Product Link http://www.matrixscientific.com/021370.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/021370.html
Availability details for
Quantity
  • Availability
  • Add to Cart