MAT100860302 Matrix Scientific

2-Cyano-3-hydroxypyridine

  • MDL number MFCD01646135
  • Cas Number : 932-35-4
  • Molecular Weight : 120.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 211-212�c.
Purity 98
Country of Origin USA
Product Link http://www.matrixscientific.com/021371.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/021371.html
Availability details for
Quantity
  • Availability
  • Add to Cart