MAT113926099 Matrix Scientific

N-[3-(2-Amino-4-chlorophenoxy)propyl]-N,N-dimethylamine

  • MDL number MFCD08686790
  • Cas Number : 1138-12-1
  • Molecular Weight : 228.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient/argon
Country of Origin USA
Product Link http://www.matrixscientific.com/021954.html

Safety Information

hcodes Irritant/Stored under Argon
MSDS http://www.matrixscientific.com/021954.html
Availability details for
Quantity
  • Availability
  • Add to Cart