MAT113926126 Matrix Scientific

N-[3-(4-Amino-2-methylphenoxy)phenyl]acetamide

  • MDL number MFCD08686822
  • Cas Number : 307308-68-5
  • Molecular Weight : 256.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/022684.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/022684.html
Availability details for
Quantity
  • Availability
  • Add to Cart