MAT113926145 Matrix Scientific

N-{3-[2-Amino-4-(trifluoromethyl)phenoxy]-phenyl}acetamide

  • MDL number MFCD08686844
  • Cas Number : 946715-45-3
  • Molecular Weight : 310.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/022062.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/022062.html
Availability details for
Quantity
  • Availability
  • Add to Cart