MAT113926159 Matrix Scientific

(2S,4S)-4-(2-Benzyl-4-chlorophenoxy)-1-(tert-butoxycarbonyl)-2-pyrrolidinecarboxylic acid

  • MDL number MFCD08686853
  • Cas Number : 1354484-99-3
  • Molecular Weight : 431.92

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/023602.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/023602.html
Availability details for
Quantity
  • Availability
  • Add to Cart