MAT113926173 Matrix Scientific

(2S,4S)-4-[4-(Benzyloxy)phenoxy]-1-(tert-butoxy-carbonyl)-2-pyrrolidinecarboxylic acid

  • MDL number MFCD08686874
  • Cas Number : 1354487-22-1
  • Molecular Weight : 413.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/023568.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/023568.html
Availability details for
Quantity
  • Availability
  • Add to Cart