MAT113926224 Matrix Scientific

(2S,4S)-4-([1,1'-Biphenyl]-4-yloxy)-1-(tert-butoxycarbonyl)-2-pyrrolidinecarboxylic acid

  • MDL number MFCD08686929
  • Cas Number : 1354485-81-6
  • Molecular Weight : 383.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/023571.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/023571.html
Availability details for
Quantity
  • Availability
  • Add to Cart