MAT113926257 Matrix Scientific

4-Bromo-2-chlorobenzotrifluoride

  • MDL number MFCD08459292
  • Cas Number : 467435-07-0
  • Molecular Weight : 259.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.matrixscientific.com/021423.html

Safety Information

hcodes Irritant/Irritant-Harmful
MSDS http://www.matrixscientific.com/021423.html
Availability details for
Quantity
  • Availability
  • Add to Cart