MAT113926266 Matrix Scientific

(2S,4S)-4-(2-Bromo-4-chlorophenoxy)-1-(tert-butoxycarbonyl)-2-pyrrolidinecarboxylic acid

  • MDL number MFCD08686969
  • Cas Number : 1354484-82-4
  • Molecular Weight : 420.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/023562.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/023562.html
Availability details for
Quantity
  • Availability
  • Add to Cart