MAT113926294 Matrix Scientific

1-Bromo-4-(ethylsulfonyl)benzene

  • MDL number MFCD08692383
  • Cas Number : 26732-20-7
  • Molecular Weight : 249.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/024313.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/024313.html
Availability details for
Quantity
  • Availability
  • Add to Cart