MAT113926301 Matrix Scientific

(2S,4S)-4-(4-Bromo-3-fluorophenoxy)-1-(tert-butoxycarbonyl)-2-pyrrolidinecarboxylic acid

  • MDL number MFCD08686999
  • Cas Number : 1354486-45-5
  • Molecular Weight : 404.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/023610.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/023610.html
Availability details for
Quantity
  • Availability
  • Add to Cart