MAT113926320 Matrix Scientific

5-Bromo-2-(3-methoxypropoxy)aniline

  • MDL number MFCD08687018
  • Cas Number : 946786-59-0
  • Molecular Weight : 260.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/022902.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/022902.html
Availability details for
Quantity
  • Availability
  • Add to Cart