MAT113926337 Matrix Scientific

5-Bromo-2-[2-(4-morpholinyl)ethoxy]aniline

  • MDL number MFCD08687034
  • Cas Number : 946786-56-7
  • Molecular Weight : 301.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 66-67
Country of Origin USA
Product Link http://www.matrixscientific.com/022898.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/022898.html
Availability details for
Quantity
  • Availability
  • Add to Cart