MAT113926351 Matrix Scientific

2-Bromo-4-(tert-pentyl)phenol

  • MDL number MFCD09055133
  • Cas Number : 18966-64-8
  • Molecular Weight : 243.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/024241.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/024241.html
Availability details for
Quantity
  • Availability
  • Add to Cart