MAT113926406 Matrix Scientific

(2S,4S)-1-(tert-Butoxycarbonyl)-4-(2,3-dihydro-1H-inden-5-yloxy)-2-pyrrolidinecarboxylic acid

  • MDL number MFCD08687097
  • Cas Number : 1354486-66-0
  • Molecular Weight : 347.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/023583.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/023583.html
Availability details for
Quantity
  • Availability
  • Add to Cart