MAT113926412 Matrix Scientific

(2S,4S)-1-(tert-Butoxycarbonyl)-4-(3,5-dimethyl-phenoxy)-2-pyrrolidinecarboxylic acid

  • MDL number MFCD08687103
  • Cas Number : 869682-05-3
  • Molecular Weight : 335.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/023557.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/023557.html
Availability details for
Quantity
  • Availability
  • Add to Cart