MAT113926418 Matrix Scientific

(2S,4S)-1-(tert-Butoxycarbonyl)-4-(3-ethyl-phenoxy)-2-pyrrolidinecarboxylic acid

  • MDL number MFCD08687109
  • Cas Number : 1354485-37-2
  • Molecular Weight : 335.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/023558.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/023558.html
Availability details for
Quantity
  • Availability
  • Add to Cart