MAT113926427 Matrix Scientific

(2S,4S)-1-(tert-Butoxycarbonyl)-4-(5-isopropyl-2-methylphenoxy)-2-pyrrolidinecarboxylic acid

  • MDL number MFCD08687118
  • Cas Number : 1354486-53-5
  • Molecular Weight : 363.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/023538.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/023538.html
Availability details for
Quantity
  • Availability
  • Add to Cart