MAT113926430 Matrix Scientific

(2S,4S)-1-(tert-Butoxycarbonyl)-4-(4-isopropyl-phenoxy)-2-pyrrolidinecarboxylic acid

  • MDL number MFCD08687121
  • Cas Number : 1354486-32-0
  • Molecular Weight : 349.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/023577.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/023577.html
Availability details for
Quantity
  • Availability
  • Add to Cart