MAT113926433 Matrix Scientific

(2S,4S)-1-(tert-Butoxycarbonyl)-4-(2-methoxy-phenoxy)-2-pyrrolidinecarboxylic acid

  • MDL number MFCD08687124
  • Cas Number : 1354486-81-9
  • Molecular Weight : 337.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/023529.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/023529.html
Availability details for
Quantity
  • Availability
  • Add to Cart