MAT113926441 Matrix Scientific

(2S,4S)-1-(tert-Butoxycarbonyl)-4-[4-(1-methyl-1-phenylethyl)phenoxy]-2-pyrrolidinecarboxylic acid

  • MDL number MFCD08687132
  • Cas Number : 1354485-42-9
  • Molecular Weight : 425.53

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/023573.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/023573.html
Availability details for
Quantity
  • Availability
  • Add to Cart