MAT113926470 Matrix Scientific

3-Butoxypyrrolidine

  • MDL number MFCD08687161
  • Cas Number : 946715-13-5
  • Molecular Weight : 143.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/023342.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/023342.html
Availability details for
Quantity
  • Availability
  • Add to Cart