MAT113926497 Matrix Scientific

4-[4-(tert-Butyl)phenoxy]-3-fluorophenylamine

  • MDL number MFCD08687188
  • Cas Number : 946785-77-9
  • Molecular Weight : 259.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/022781.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/022781.html
Availability details for
Quantity
  • Availability
  • Add to Cart