MAT113926524 Matrix Scientific

4-[3-(tert-Butyl)phenoxy]-3-(trifluoromethyl)-phenylamine

  • MDL number MFCD08687214
  • Cas Number : 946740-74-5
  • Molecular Weight : 309.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/022528.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/022528.html
Availability details for
Quantity
  • Availability
  • Add to Cart