MAT113926594 Matrix Scientific

3-Chloro-4-(2-ethoxyphenoxy)aniline

  • MDL number MFCD08687296
  • Cas Number : 89013-43-4
  • Molecular Weight : 263.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/022365.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/022365.html
Availability details for
Quantity
  • Availability
  • Add to Cart