MAT113926742 Matrix Scientific

2-(Allyloxy)benzohydrazide

  • MDL number MFCD06447101
  • Cas Number : 18167-41-4
  • Molecular Weight : 192.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/024016.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/024016.html
Availability details for
Quantity
  • Availability
  • Add to Cart