MAT113926765 Matrix Scientific

(2S,4S)-1-(tert-Butoxycarbonyl)-4-(2-naphthyloxy)-2-pyrrolidinecarboxylic acid

  • MDL number MFCD06656449
  • Cas Number : 317357-42-9
  • Molecular Weight : 357.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/023586.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/023586.html
Availability details for
Quantity
  • Availability
  • Add to Cart