MAT113926766 Matrix Scientific

4-[([1,1'-Biphenyl]-2-yloxy)methyl]piperidine

  • MDL number MFCD06245983
  • Cas Number : 883514-41-8
  • Molecular Weight : 267.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/023085.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/023085.html
Availability details for
Quantity
  • Availability
  • Add to Cart