MAT113926783 Matrix Scientific

3,5-Bis(trifluoromethyl)phenyldimethylchlorosilane

  • MDL number MFCD01862186
  • Cas Number : 732306-23-9
  • Molecular Weight : 306.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Boiling point 95-96�mm
Product Link http://www.matrixscientific.com/023808.html

Safety Information

hcodes Corrosive/Moisture Sensitive
MSDS http://www.matrixscientific.com/023808.html
Availability details for
Quantity
  • Availability
  • Add to Cart