MAT113926792 Matrix Scientific

3-Bromo-4-butoxybenzoic acid

  • MDL number MFCD06662499
  • Cas Number : 450416-09-8
  • Molecular Weight : 273.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/024092.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/024092.html
Availability details for
Quantity
  • Availability
  • Add to Cart