MAT113926834 Matrix Scientific

2-(2-Bromo-4-chlorophenoxy)acetic acid

  • MDL number MFCD03613709
  • Cas Number : 77228-66-1
  • Molecular Weight : 265.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/024028.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/024028.html
Availability details for
Quantity
  • Availability
  • Add to Cart