MAT113926848 Matrix Scientific

3-Amino-4-(trifluoromethyl)benzoic acid

  • MDL number MFCD07784307
  • Cas Number : 125483-00-3
  • Molecular Weight : 205.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 203-205
Purity 98
Country of Origin USA
Product Link http://www.matrixscientific.com/021433.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/021433.html
Availability details for
Quantity
  • Availability
  • Add to Cart