MAT113926973 Matrix Scientific

3-Bromo-4-isopropoxybenzoic acid

  • MDL number MFCD07187811
  • Cas Number : 213598-20-0
  • Molecular Weight : 259.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/024110.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/024110.html
Availability details for
Quantity
  • Availability
  • Add to Cart