MAT113927002 Matrix Scientific

2-Chloro-8-ethyl-quinoline-3-carbaldehyde

  • MDL number MFCD02587932
  • Cas Number : 335196-05-9
  • Molecular Weight : 219.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/021444.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/021444.html
Availability details for
Quantity
  • Availability
  • Add to Cart