MAT113927003 Matrix Scientific

2-Chloro-6-fluoroaniline

  • MDL number MFCD00040309
  • Cas Number : 363-51-9
  • Molecular Weight : 145.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 31-32
Purity 97
Density 1.316/25
Country of Origin USA
Boiling point 67�mm
Product Link http://www.matrixscientific.com/002765.html

Safety Information

hcodes Toxic
MSDS http://www.matrixscientific.com/002765.html
Availability details for
Quantity
  • Availability
  • Add to Cart