MAT113927005 Matrix Scientific

5-Chloro-2-fluorobenzenesulfonamide

  • MDL number MFCD03094289
  • Cas Number : 351003-57-1
  • Molecular Weight : 209.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 134-136
Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/021394.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/021394.html
Availability details for
Quantity
  • Availability
  • Add to Cart