MAT113927021 Matrix Scientific

2-(2-Bromo-4-methylphenoxy)acetic acid

  • MDL number MFCD00029787
  • Cas Number : 25181-66-2
  • Molecular Weight : 245.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/024046.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/024046.html
Availability details for
Quantity
  • Availability
  • Add to Cart