MAT113927029 Matrix Scientific

N-Benzyl(cyclopropyl)methanamine

  • MDL number MFCD03129358
  • Cas Number : 116373-23-0
  • Molecular Weight : 161.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/024205.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/024205.html
Availability details for
Quantity
  • Availability
  • Add to Cart