MAT113927033 Matrix Scientific

N-Benzyl(3-methylphenyl)methanamine hydrochloride

  • MDL number MFCD03930957
  • Cas Number : 15429-17-1
  • Molecular Weight : 247.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/024147.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/024147.html
Availability details for
Quantity
  • Availability
  • Add to Cart