MAT113927596 Matrix Scientific

5-Chloro-2-(2-methoxyphenoxy)phenylamine

  • MDL number MFCD08687367
  • Cas Number : 946682-52-6
  • Molecular Weight : 249.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/021973.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/021973.html
Availability details for
Quantity
  • Availability
  • Add to Cart